1-ETHYL-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIO 1 * 250 mg
Catalog # APOSOR933536-250MG
Supplier: Apollo Scientific
1-ETHYL-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIO 1 * 250 mg
Catalog # APOSOR933536-250MG
Supplier: Apollo Scientific
Frequently Bought Together
1-Butanol, EMPROVE® ESSENTIAL NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetonitrile, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2-Propanol ≥99.9%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol absolute 99.8% for analysis
Supplier: Thermo Fisher Scientific
Ethanol ≥96% (v/v)
Supplier: KLINIPATH
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane ACS, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
Methanol 99.9% for analysis
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Nitrobenzene 99+% ACS
Supplier: Thermo Fisher Scientific