TIP FITTING 0.1-10?L F1-CT MICRO 1 * 1 items
Catalog # LABS1067750-VAN
Supplier: Thermo Fisher Scientific
TIP FITTING 0.1-10?L F1-CT MICRO 1 * 1 items
Catalog # LABS1067750-VAN
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Grease with stick 1 * 1 items
Supplier: EPPENDORF
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
SONO-CHECK 1 * 30 items
Supplier: KING DISPOSABLES
Accessories for S1 pipette controller
Supplier: Thermo Fisher Scientific
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Overpressur plug 1 * 1 items
Supplier: VWR Collection
Accessories for Titrette®
Supplier: Brand
control cable, Motor 82430M-S02 - control board V2S2 1 * 1 items
Supplier: GRANT INSTRUMENTS
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Rotors and accessories for Megafuge® 8 centrifuges
Supplier: Thermo Scientific
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
SHAFT KIT UHP 10 W/SEAL+ORING 1 * 1 KIT
Supplier: VWR Collection
Steam release valve 1 * 1 items
Supplier: CERTOCLAV
CONNECTION VACUUM 1 * 1 items
Supplier: IKA
FELT RING 1 * 1 items
Supplier: IKA
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach