Order Entry
Netherlands
ContactUsLinkComponent
Rapidly identify Staphylococcus aureus and distinguish from other staphylococci by detecting clumping factor and protein A using the Thermo Scientific BactiStaph Latex Agglutination Test Kit. 1 * 1 KIT
Catalog # OXOIR21144
Rapidly identify Staphylococcus aureus and distinguish from other staphylococci by detecting clumping factor and protein A using the Thermo Scientific BactiStaph Latex Agglutination Test Kit. 1 * 1 KIT
Catalog # OXOIR21144
Frequently Bought Together