2-Bromo-5-methylthiophene,98% (stabilized with Copper chip and NaHCO3) 1 * 10 g
Catalog # AMBDA100942-10G
Supplier: AMBEED
2-Bromo-5-methylthiophene,98% (stabilized with Copper chip and NaHCO3) 1 * 10 g
Catalog # AMBDA100942-10G
Supplier: AMBEED
Frequently Bought Together
Diethyl ether EMPROVE ESSENTIAL Ph Eur,BP 1 * 190 L
Supplier: MERCK PRODUCTION CHEMICALS
N,N-Dimethylformamide ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Phosphorus(III) chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
ACETONE PURE PHARMA GRADE 1 * 200 L
Supplier: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Water, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Cyclohexane 99+% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.5% NF, Ph. Eur., BP
Supplier: VWR Chemicals