tert-Butyl (2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl) carbonate,98% 1 * 0,25 g
Catalog # AMBDA333604-250MG
Supplier: AMBEED
tert-Butyl (2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl) carbonate,98% 1 * 0,25 g
Catalog # AMBDA333604-250MG
Supplier: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals