2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol,98% 1 * 0,25 g
AMBDA787735-250MG
: AMBEED
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol,98% 1 * 0,25 g
AMBDA787735-250MG
: AMBEED
Frequently Bought Together
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals