2-(3-Fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,1 g
AMBDA196195-100MG
: AMBEED
2-(3-Fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,1 g
AMBDA196195-100MG
: AMBEED
Frequently Bought Together
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
: Thermo Fisher Scientific
Ethylenediamine 99%
: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Toluene 99.5% ACS for spectroscopy
: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
: PanReac AppliChem
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
: Thermo Fisher Scientific