2-(5-Bromo-2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,97% 1 * 0,25 g
AMBDA707176-250MG
: AMBEED
2-(5-Bromo-2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,97% 1 * 0,25 g
AMBDA707176-250MG
: AMBEED
Frequently Bought Together
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
: VWR Chemicals
Glycerine 85% Ph. Eur.
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals