2-(2-(Bromomethyl)-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,1 g
AMBDA910747-100MG
: AMBEED
2-(2-(Bromomethyl)-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,1 g
AMBDA910747-100MG
: AMBEED
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals