2-(4-((Cyclopentylthio)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA281252-250MG
Supplier: AMBEED
2-(4-((Cyclopentylthio)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA281252-250MG
Supplier: AMBEED
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
p-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol, dehydrated (max. 0.01% H₂O) ≥99.8%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals