(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-fluoro-1H-indol-3-yl)propanoic acid,95% 1 * 0,1 g
Catalog # AMBDA210577-100MG
Supplier: AMBEED
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-fluoro-1H-indol-3-yl)propanoic acid,95% 1 * 0,1 g
Catalog # AMBDA210577-100MG
Supplier: AMBEED
Frequently Bought Together
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: Honeywell Chemicals
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem