2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA296941-250MG
Supplier: AMBEED
2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA296941-250MG
Supplier: AMBEED
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
1-Butanol NF
Supplier: Spectrum Chemical
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific