(3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol,98% 1 * 0,25 g
Catalog # AMBDA657949-250MG
Supplier: AMBEED
(3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol,98% 1 * 0,25 g
Catalog # AMBDA657949-250MG
Supplier: AMBEED
Frequently Bought Together
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butanol ≥99%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Carbon disulphide ≥99.9%, Reagent Grade low in benzene
Supplier: Honeywell Chemicals