N-Methyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine,98% 1 * 0,1 g
Catalog # AMBDA319193-100MG
Supplier: AMBEED
N-Methyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine,98% 1 * 0,1 g
Catalog # AMBDA319193-100MG
Supplier: AMBEED
Frequently Bought Together
o-Xylene anhydrous, 97 % 1 * 1 L
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific