4-Methoxy-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylidene)aniline,98% 1 * 0,25 g
Catalog # AMBDA163447-250MG
Supplier: AMBEED
4-Methoxy-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylidene)aniline,98% 1 * 0,25 g
Catalog # AMBDA163447-250MG
Supplier: AMBEED
Frequently Bought Together
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Dimethyl sulfoxide anhydrous, >= 99.9 % 1 * 2 L
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water for LC-MS 1 * 2,5 L
Supplier: PanReac AppliChem
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Glycerine ≥99%
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific