N-((Tetrahydrofuran-2-yl)methyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide,97% 1 * 0,25 g
AMBDA515127-250MG
: AMBEED
N-((Tetrahydrofuran-2-yl)methyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide,97% 1 * 0,25 g
AMBDA515127-250MG
: AMBEED
Frequently Bought Together
Cyclohexane for pesticide residue analysis
: Thermo Fisher Scientific
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
: PanReac AppliChem
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Nitrobenzene 99+% ACS
: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals
Water, Ultrapure for HPLC
: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
: Honeywell Chemicals