2-(4-Chloro-2-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98+% 1 * 0,25 g
AMBDA751188-250MG
: AMBEED
2-(4-Chloro-2-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98+% 1 * 0,25 g
AMBDA751188-250MG
: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals