1-Isobutyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea,98% 1 * 0,25 g
AMBDA525488-250MG
: AMBEED
1-Isobutyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea,98% 1 * 0,25 g
AMBDA525488-250MG
: AMBEED
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
: Merck
Water, PESTINORM® for headspace gas chromatography
: VWR Chemicals
N,N-Dimethylacetamide 99%, pure
: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol, extra dry 99.8%, AcroSeal®
: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
: Thermo Fisher Scientific