N-(4-Methoxybenzyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine,98% 1 * 0,1 g
Catalog # AMBDA229535-100MG
Supplier: AMBEED
N-(4-Methoxybenzyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine,98% 1 * 0,1 g
Catalog # AMBDA229535-100MG
Supplier: AMBEED
Frequently Bought Together
Turpentine oil, TECHNICAL
Supplier: VWR Chemicals
Hexane anhydrous, 95 % 1 * 250 mL
Supplier: Merck
Trifluoroacetic acid ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
1-Propanol, EMPLURA®, Supelco®
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform stabilised for HPLC
Supplier: Thermo Fisher Scientific
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific