4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde,98% 1 * 0,25 g
Catalog # AMBDA141600-250MG
Supplier: AMBEED
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde,98% 1 * 0,25 g
Catalog # AMBDA141600-250MG
Supplier: AMBEED
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals