2,2,2-TRIFLUORO-1-(3-(4,4,5,5-TETRAMETHY 1 * 0,25 g
Catalog # AMBDA418525-250MG
Supplier: AMBEED
Frequently Bought Together
Dimethylsulfoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Chloroform, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
[EN]1,4-DIOXANE ANHYDROUS, 99.8 %, CONTA 1 * 1 L
Supplier: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitril 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylacetaat ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute (untaxed) ≥99.8%, Puriss. p.a. ACS, A15, without additive, Riedel-de Haën™
Supplier: Honeywell Chemicals
PETROLEUM ETHER 40-60C FOR PESTICIDE AN 1 * 2,5 L
Supplier: PanReac AppliChem
Aceton, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals