2-(5-Chloro-4-methylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,96% 1 * 0,1 g
Catalog # AMBDA213401-100MG
Supplier: AMBEED
2-(5-Chloro-4-methylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,96% 1 * 0,1 g
Catalog # AMBDA213401-100MG
Supplier: AMBEED
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.8% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Water for analysis, ACS 1 * 5 L
Supplier: PanReac AppliChem
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Tetramethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals