(E)-2-(4-Chlorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98+% 1 * 0,25 g
Catalogus # AMBDA914363-250MG
Supplier: AMBEED
(E)-2-(4-Chlorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98+% 1 * 0,25 g
Catalogus # AMBDA914363-250MG
Supplier: AMBEED
Frequently Bought Together
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetone, EMPLURA®, Reagent Grade for synthesis, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Ethanol absolute (untaxed) ≥99.8%, Puriss. p.a. ACS, A15, without additive, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Methyl ethyl ketone, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
Supplier: Honeywell Chemicals