4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]decan-8-yl)-1,3,2-dioxaborolane,95% 1 * 0,25 g
Catalog # AMBDA718116-250MG
Supplier: AMBEED
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]decan-8-yl)-1,3,2-dioxaborolane,95% 1 * 0,25 g
Catalog # AMBDA718116-250MG
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP) ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8% ACS
Supplier: Thermo Fisher Scientific