1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole,98% 1 * 0,25 g
Catalog # AMBDA871312-250MG
Supplier: AMBEED
1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole,98% 1 * 0,25 g
Catalog # AMBDA871312-250MG
Supplier: AMBEED
Frequently Bought Together
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzoic acid methyl ester
Supplier: Merck
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Anisole, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific