Ethyl 2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetate,98% 1 * 0,1 g
Catalog # AMBDA441366-100MG
Supplier: AMBEED
Ethyl 2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetate,98% 1 * 0,1 g
Catalog # AMBDA441366-100MG
Supplier: AMBEED
Frequently Bought Together
Methyl acetate anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
(±)-2-Ethyl-1-hexanol ≥99%
Supplier: Thermo Fisher Scientific
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
TOLUOL CHROMASOLV FUER DI 1 * 18 L
Supplier: Honeywell Chemicals
2-Propanol ≥99.7% for gas chromatography
Supplier: Thermo Fisher Scientific
1,4-Dioxane, extra dry 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Propanol 99+%, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals