2-(5-Chloro-2-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,95% 1 * 0,25 g
Catalog # AMBDA1627612-250MG
Supplier: AMBEED
2-(5-Chloro-2-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,95% 1 * 0,25 g
Catalog # AMBDA1627612-250MG
Supplier: AMBEED
Frequently Bought Together
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
(±)-2-Butanol ≥99% analytical reagent
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Diethyl ether, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol 99.5+%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals