N,N-Diethyl-3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide,98% 1 * 0,25 g
Catalog # AMBDA1624495-250MG
Supplier: AMBEED
N,N-Diethyl-3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide,98% 1 * 0,25 g
Catalog # AMBDA1624495-250MG
Supplier: AMBEED
Frequently Bought Together
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetone, J.T.Baker®
Supplier: Avantor
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform stabilized with 50 ppm of amylene (Reag. USP, Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific