2-(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,95% 1 * 1 g
Catalog # AMBDA1627830-1G
Supplier: AMBEED
2-(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,95% 1 * 1 g
Catalog # AMBDA1627830-1G
Supplier: AMBEED
Frequently Bought Together
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Methylpyrrolidine 98%
Supplier: Thermo Fisher Scientific
Diiodomethane 99+% stabilised
Supplier: Thermo Fisher Scientific
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99%
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor