(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,25 g
Catalog # AMBDA1636066-250MG
Supplier: AMBEED
(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,25 g
Catalog # AMBDA1636066-250MG
Supplier: AMBEED
Frequently Bought Together
Hexane anhydrous, 95 % 1 * 250 mL
Supplier: Merck
1-Butanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethanol (untaxed) ~96% (v/v), CHROMASOLV™ ACS for UV spectrophotometry extra fine spirit, E15 o 1, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
2-Propanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem