Ethyl 4-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate,95+% 1 * 0,1 g
AMBDA1538299-100MG
: AMBEED
Ethyl 4-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate,95+% 1 * 0,1 g
AMBDA1538299-100MG
: AMBEED
Frequently Bought Together
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
: PanReac AppliChem
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Cyclohexane for pesticide residue analysis
: Thermo Fisher Scientific
(±)-2-Ethyl-1-hexanol 99%
: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Amyl Alcohol according to NF V 04-210 for analysis 1 * 1.000 mL
: PanReac AppliChem
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
: Honeywell Chemicals
Methanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Diethyl ether stabilised ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
: Honeywell Chemicals