4,4,5,5-Tetramethyl-2-(5-nitronaphthalen-1-yl)-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA1194680-250MG
Supplier: AMBEED
4,4,5,5-Tetramethyl-2-(5-nitronaphthalen-1-yl)-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA1194680-250MG
Supplier: AMBEED
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Toluene ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals