(E)-tert-Butyldimethyl((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)allyl)oxy)silane,95% 1 * 0,25 g
Catalog # AMBDA1164792-250MG
Supplier: AMBEED
(E)-tert-Butyldimethyl((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)allyl)oxy)silane,95% 1 * 0,25 g
Catalog # AMBDA1164792-250MG
Supplier: AMBEED
Frequently Bought Together
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific