4-Bromo-1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-indole,95% 1 * 0,25 g
Catalog # AMBDA730112-250MG
Supplier: AMBEED
4-Bromo-1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-indole,95% 1 * 0,25 g
Catalog # AMBDA730112-250MG
Supplier: AMBEED
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific