Methyl 2-bromo-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate,98% 1 * 0,25 g
Catalog # AMBDA969849-250MG
Leverancier: AMBEED
Methyl 2-bromo-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate,98% 1 * 0,25 g
Catalog # AMBDA969849-250MG
Leverancier: AMBEED
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
Leverancier: VWR Chemicals
n-Hexane ≥98%, SupraSolv® for GC-MS
Leverancier: Merck
Water, TECHNICAL, demineralised
Leverancier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Methanol, EMPLURA®, Supelco®
Leverancier: Merck
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Leverancier: Merck
Isoamyl alcohol, TECHNICAL
Leverancier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Leverancier: Merck
Petroleum spirit, 40…60 °C, EMSURE® ACS, ISO for analysis, Supelco®
Leverancier: Merck
Dimethyl sulphoxide, dehydrated (max. 0.03% H₂O) ≥99.9%, AnalaR NORMAPUR® ACS analytical reagent
Leverancier: VWR Chemicals
Ethanol absolute ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Toluene, EMPLURA®, extra pure, Supelco®
Leverancier: Merck
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Leverancier: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals
n-Heptane
Leverancier: Merck
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Leverancier: Thermo Fisher Scientific