2-(2-(Dibromomethyl)-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,97% 1 * 0,1 g
AMBDA922071-100MG
: AMBEED
2-(2-(Dibromomethyl)-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,97% 1 * 0,1 g
AMBDA922071-100MG
: AMBEED
Frequently Bought Together
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol 99.5+%
: Thermo Fisher Scientific