(2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-((E)-4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol,98% 1 * 0,1 g
AMBDA263851-100MG
: AMBEED
(2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-((E)-4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol,98% 1 * 0,1 g
AMBDA263851-100MG
: AMBEED
Frequently Bought Together
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.0% USP, Ph. Eur.
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Tetrabutylammonium dihydrogen phosphate 0.5 M in HPLC grade water for HPLC
: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific
n-Heptane 95%
: Thermo Fisher Scientific
Acetonitrile 99+% for spectroscopy
: Thermo Fisher Scientific