4-Amino-1-((2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one,95% 1 * 5 g
Catalog # AMBDA148637-5G
Supplier: AMBEED
4-Amino-1-((2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one,95% 1 * 5 g
Catalog # AMBDA148637-5G
Supplier: AMBEED
Frequently Bought Together
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Hydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane
Supplier: Roth Carl
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Acetone, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific