2-Chloronaphthalene-1,4-dione,97% 1 * 10 g
Catalog # AMBDA145307-10G
Supplier: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol 99.5%, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.9+% ACS
Supplier: Thermo Fisher Scientific
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
1,4-Dioxane, extra dry 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific