Triethoxy(3-(oxiran-2-ylmethoxy)propyl)silane,98% +(GC) 1 * 25 g
Catalog # AMBDA179895-25G
Supplier: AMBEED
Triethoxy(3-(oxiran-2-ylmethoxy)propyl)silane,98% +(GC) 1 * 25 g
Catalog # AMBDA179895-25G
Supplier: AMBEED
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific