2-Methoxyethyl methacrylate,98% (stabilized with MEHQ) 1 * 25 g
Catalog # AMBDA547163-25G
Leverancier: AMBEED
2-Methoxyethyl methacrylate,98% (stabilized with MEHQ) 1 * 25 g
Catalog # AMBDA547163-25G
Leverancier: AMBEED
Frequently Bought Together
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Leverancier: Honeywell Chemicals
Isohexane (mixture of isomers), EMSURE® for analysis, Supelco®
Leverancier: Merck
1-Propanol, EMPLURA®, Supelco®
Leverancier: Merck
Glycerine 85% Ph. Eur.
Leverancier: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Leverancier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Leverancier: VWR Chemicals
Methanol, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Leverancier: Avantor
Water, BAKER ANALYZED® for LC-MS, J.T.Baker®
Leverancier: Avantor
n-Heptane, anhydrous 99+%
Leverancier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9+%, AcroSeal®
Leverancier: Thermo Fisher Scientific
p-Xylene 99%, pure
Leverancier: Thermo Fisher Scientific
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Benzyl alcohol ≥99%
Leverancier: Thermo Fisher Scientific
1,4-Dioxane, anhydrous ≥99.8%
Leverancier: Thermo Fisher Scientific
Carbon disulphide 99.9%, extra pure low in benzene
Leverancier: Thermo Fisher Scientific