2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide,95% 1 * 10 g
Catalog # AMBDA237594-10G
Supplier: AMBEED
2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide,95% 1 * 10 g
Catalog # AMBDA237594-10G
Supplier: AMBEED
Frequently Bought Together
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Ethyl-1-hexanol 99%
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals