3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate,99% 1 * 5 g
AMBDA367713-5G
: AMBEED
3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate,99% 1 * 5 g
AMBDA367713-5G
: AMBEED
Frequently Bought Together
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
: Thermo Fisher Scientific
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
n-Hexane 95+%, extra pure
: Thermo Fisher Scientific
Chlorobenzene ≥99%
: Thermo Fisher Scientific
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
: Honeywell Chemicals
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
: VWR Chemicals
Pyridine ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals
Cesium fluoride 99.9%, extra pure
: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal®
: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
: Honeywell Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
: Thermo Fisher Scientific