N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide,98% 1 * 1.000 g
Catalog # AMBDA258344-1000G
Supplier: AMBEED
N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide,98% 1 * 1.000 g
Catalog # AMBDA258344-1000G
Supplier: AMBEED
Frequently Bought Together
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem