5-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine,98% 1 * 0,25 g
Catalog # AMBDA871057-250MG
Supplier: AMBEED
5-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine,98% 1 * 0,25 g
Catalog # AMBDA871057-250MG
Supplier: AMBEED
Frequently Bought Together
tert-Butyl methyl ether ≥99.0% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor
Methanol ≥99.8% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals