Potassium trifluoro(1,4-dioxaspiro[4.5]dec-7-en-8-yl)borate,98% 1 * 0,25 g
AMBDA820412-250MG
: AMBEED
Potassium trifluoro(1,4-dioxaspiro[4.5]dec-7-en-8-yl)borate,98% 1 * 0,25 g
AMBDA820412-250MG
: AMBEED
Frequently Bought Together
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Diethyl ether stabilised Ph. Eur.
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
: VWR Chemicals
Glycerine, Hi-LR™
: HIMEDIA
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
: Thermo Fisher Scientific