N-(Trifluoromethylthio)phthalimide,98% 1 * 5 g
Catalog # AMBDA442652-5G
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Allyl alcohol for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane, BAKER ANALYZED® HPLC for HPLC, J.T.Baker®
Supplier: Avantor
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
1,2,3,4-TETRAHYDRONAPHTHALENE FOR SYN... 1 * 200 kg
Supplier: Merck
Pyridine, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Mineral oil (petroleum) for IR spectrophotometry, suitable for nujol mull preparations
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) ≥99.8%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Ethanol ≥96% (v/v)
Supplier: KLINIPATH
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals