1-Methyl-4-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)piperazine,96% 1 * 0,1 g
Catalog # AMBDA727022-100MG
Supplier: AMBEED
1-Methyl-4-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)piperazine,96% 1 * 0,1 g
Catalog # AMBDA727022-100MG
Supplier: AMBEED
Frequently Bought Together
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
1-Propanol, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals