2-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethoxy)acetic acid,97% 1 * 5 g
Catalog # AMBDA166606-5G
Supplier: AMBEED
2-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethoxy)acetic acid,97% 1 * 5 g
Catalog # AMBDA166606-5G
Supplier: AMBEED
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1 L
Supplier: PanReac AppliChem
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Butanone (Methylethylketone) (Reag. USP, Ph. Eur.) for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem