Ethene-1,1-diyldibenzene,98% (stabilized with HQ) 1 * 25 g
Catalog # AMBDA104720-25G
Supplier: AMBEED
Ethene-1,1-diyldibenzene,98% (stabilized with HQ) 1 * 25 g
Catalog # AMBDA104720-25G
Supplier: AMBEED
Frequently Bought Together
1-Butanol, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.0% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Dichloromethane ≥99.9% stabilised, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Pyridine, EMPARTA® ACS analytical reagent
Supplier: Merck
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol absolute 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals